Learn More
Medchemexpress LLC Trans-Benzylideneacetone | 1896-62-4 | 99.87% | 146.19 | 1 G
SDP
Catalog No. 5000366578
Encompass_Preferred
trans-Benzylideneacetone (trans-Benzalacetone) is a metabolite of the gram-negative entomopathogenic bacterium *Xenorhabdus nematophila*. It acts as an enzyme inhibitor against phospholipase A2 (PLA2) and is also an immunosuppressant, intended for research use only.
  • High purity of 99.87%.
  • Inhibits phospholipase A2 (PLA2) activity.
  • Acts as an immunosuppressant.
  • Suitable for various *in vitro* and *in vivo* research protocols.
  • Store at 4°C, protected from light.
Catalog No. 50-003-66578 Supplier Medchemexpress LLC Supplier No. HYW012595A1G
trans-Benzylideneacetone (trans-Benzalacetone) is a metabolite of the gram-negative entomopathogenic bacterium *Xenorhabdus nematophila*. It acts as an enzyme inhibitor against phospholipase A2 (PLA2) and is also an immunosuppressant, intended for research use only.
  • High purity of 99.87%.
  • Inhibits phospholipase A2 (PLA2) activity.
  • Acts as an immunosuppressant.
  • Suitable for various *in vitro* and *in vivo* research protocols.
  • Store at 4°C, protected from light.