Learn More
Medchemexpress LLC Trans-Benzylideneacetone | 1896-62-4 | 99.87% | 146.19 | 10 G
SDP
Catalog No. 5000453872
Encompass_Preferred
trans-Benzylideneacetone (trans-Benzalacetone) is a metabolite of gram-negative entomopathogenic bacterium *Xenorhabdus nematophila*, serving as an enzyme inhibitor against phospholipase A2 (PLA2). It is also an immunosuppressant.
  • Inhibits hemocyte-spreading behavior in *S. exigua* larvae
  • Enhances the virulence of *Bacillus thuringiensis* against beet armyworm
  • Suitable for research on enzyme inhibition and immunosuppression
Catalog No. 50-004-53872 Supplier Medchemexpress LLC Supplier No. HYW012595A10G
trans-Benzylideneacetone (trans-Benzalacetone) is a metabolite of gram-negative entomopathogenic bacterium *Xenorhabdus nematophila*, serving as an enzyme inhibitor against phospholipase A2 (PLA2). It is also an immunosuppressant.
  • Inhibits hemocyte-spreading behavior in *S. exigua* larvae
  • Enhances the virulence of *Bacillus thuringiensis* against beet armyworm
  • Suitable for research on enzyme inhibition and immunosuppression