Learn More
Medchemexpress LLC HY-W012595A 500mg Medchemexpress, trans-Benzylideneacetone CAS:1896-62-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW012595A500MG
Encompass_Preferred
Medchemexpress, HY-W012595A 500mg trans-Benzylideneacetone CAS:1896-62-4 trans-Benzylideneacetone (trans-Benzalacetone), a metabolite of gram-negative entomopathogenic bacterium Xenorhabdus nematophila, is an enzyme inhibitor against phospholipase A2 (PLA2). trans-Benzylideneacetone is an immunosuppressant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3331