Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich trans-beta-Methylstyrene

Catalog No. 1118481G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-184-81G 1 g
11-184-810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-184-81G Supplier Sigma Aldrich Supplier No. 1118481G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The product exhibits potential exothermic hazards at higher temperatures.Trans-α-methylstyrene was used in the preparation of exo-chromans by the oxa analogue of the Povarov reaction.

Specifications

CAS 873-66-5
Density 0.911 g/mL (at 25°C (literature))
Boiling Point 175°C (lit.)
Molecular Formula C9H10
Refractive Index n20/D 1.550 (literature)
Linear Formula C6H5CH=CHCH3
MDL Number MFCD00009280
Quantity 1 g
Synonym trans-1-Phenyl-1-propene; trans-beta-Methylstyrene; trans-Propenylbenzene
Molecular Weight (g/mol) 118.18
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.