Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Rad Laboratories TRANS-BLOT TURBO RTA MINI 0.2

Catalog No. NC2387144
Encompass

  • Ready-to-assemble format
  • Nitrocellulose membrane type
  • Membrane pore size: 0.2 µm
  • Gel size compatibility: Mini
  • Includes 40 blots per kit
  • Membrane dimensions: 7 x 8.5 cm
  • Includes 40 mini-sized 0.2 µm nitrocellulose membranes
  • Includes 80 transfer stacks
  • Includes 1 L of 5x transfer buffer
  • Includes 2 gel trays
  • Compatible with Trans-Blot Turbo Transfer System
  • Application: Western blotting
  • Primary benefit: Fast protein transfer
  • Secondary benefit: High transfer efficiency
  • Cost benefit: Economical option for high-throughput labs
  • Status: Discontinued

Catalog No. NC2387144 Supplier Bio Rad Laboratories Supplier No. 1704270
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Ready-to-assemble format
  • Nitrocellulose membrane type
  • Membrane pore size: 0.2 µm
  • Gel size compatibility: Mini
  • Includes 40 blots per kit
  • Membrane dimensions: 7 x 8.5 cm
  • Includes 40 mini-sized 0.2 µm nitrocellulose membranes
  • Includes 80 transfer stacks
  • Includes 1 L of 5x transfer buffer
  • Includes 2 gel trays
  • Compatible with Trans-Blot Turbo Transfer System
  • Application: Western blotting
  • Primary benefit: Fast protein transfer
  • Secondary benefit: High transfer efficiency
  • Cost benefit: Economical option for high-throughput labs
  • Status: Discontinued

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.