Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Rad Laboratories TRANS-BLOT TURBO RTAMINI 0.2UM

Catalog No. NC1837414
Encompass

  • Product type: Transfer kit
  • Membrane type: PVDF
  • Membrane pore size: 0.2 µm
  • Gel size: Mini
  • Number of transfers: 40
  • Assembly: Requires assembly
  • Membrane dimensions: 7 cm x 8.5 cm
  • Included components: 40 mini-sized PVDF membranes, 80 transfer stacks, 1 L 5x transfer buffer, 2 gel trays
  • Application: Western blotting, protein transfer
  • System compatibility: Bio-Rad Trans-Blot Turbo Transfer System
  • Benefit: Economical option
  • Feature: Optimized buffer, membrane, and filter paper combination
  • Feature: Reduces nonspecific binding

Catalog No. NC1837414 Supplier Bio Rad Laboratories Supplier No. 1704272
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Product type: Transfer kit
  • Membrane type: PVDF
  • Membrane pore size: 0.2 µm
  • Gel size: Mini
  • Number of transfers: 40
  • Assembly: Requires assembly
  • Membrane dimensions: 7 cm x 8.5 cm
  • Included components: 40 mini-sized PVDF membranes, 80 transfer stacks, 1 L 5x transfer buffer, 2 gel trays
  • Application: Western blotting, protein transfer
  • System compatibility: Bio-Rad Trans-Blot Turbo Transfer System
  • Benefit: Economical option
  • Feature: Optimized buffer, membrane, and filter paper combination
  • Feature: Reduces nonspecific binding

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.