Learn More
Bio Rad Laboratories TRANS-BLOT TURBO5X TRNS BUFFER
Catalog No. NC3449272
Encompass
  • Concentrated solution.
  • 5x concentration.
  • 1 L volume.
  • Compatible with Trans-Blot Turbo Transfer System.
  • Application: Western blotting, protein transfer.
  • Throughput for up to 40 mini or midi-gel transfers.
  • Transfer time as quick as 3–7 minutes.
  • Membrane compatibility: PVDF, nitrocellulose.
  • Features optimized buffer and economical option.
  • Preparation requires dilution and potential additions for 1x working solution.
  • Ensures uniform protein migration.
  • High protein resolution.
  • Consistent protein transfer.
  • Superior protein transfer efficiency.
Catalog No. NC3449272 Supplier Bio Rad Laboratories Supplier No. 10026938
  • Concentrated solution.
  • 5x concentration.
  • 1 L volume.
  • Compatible with Trans-Blot Turbo Transfer System.
  • Application: Western blotting, protein transfer.
  • Throughput for up to 40 mini or midi-gel transfers.
  • Transfer time as quick as 3–7 minutes.
  • Membrane compatibility: PVDF, nitrocellulose.
  • Features optimized buffer and economical option.
  • Preparation requires dilution and potential additions for 1x working solution.
  • Ensures uniform protein migration.
  • High protein resolution.
  • Consistent protein transfer.
  • Superior protein transfer efficiency.