Learn More
Thomson Instrument Company Inversion Transfer Cap, 7/16" Male Connection, Sterile
SDP
Supplier:  Thomson Instrument Company 9317064
Encompass_Preferred
Thomson’s Inversion Transfer Caps provide a fast, aseptic, and pump-free solution for transferring culture media and reagents directly from Optimum Growth flasks into bioreactors, cell bags, or other vessels.
•Eliminate the need for intermediate bottles—connect and invert directly into wave bags, bioreactors, or other receiving vessels
•Sterile, closed system simplifies aseptic workflows—no pumps or laminar hood steps required
•Ideal for PEI-DNA complex transfers or other workflows where pumping may be too aggressive
•Reduce cell loss by up to 20% compared to traditional pump systems
•Fast setup—assembly takes approximately 5 minutes per flask
•Multiple connection options support flexible integration into various systems
•Compatible with 1.6L, 2.8L, and 5L Thomson Optimum Growth Flasks
•Gravity-driven system eliminates the need for power or pump validation steps
•Made in the USA and individually packaged for cleanroom-ready use
Catalog No. 50-111-9996