Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (-)-Trans-Caryophyllene

Catalog No. 2207525MLF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-075-25MLF 25 mL
22-075-1MLF 1 mL
22-075-5MLF 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 22-075-25MLF Supplier Sigma Aldrich Supplier No. 2207525MLF
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(-)-trans-Caryophyllene, a sesquiterpene, is the major constituent in the essential oil of Mentha longifolia and Commiphora gileadensis. It shows anti-inflammatory, local anaesthetic and antifungal properties. (-)-trans-Caryophyllene also shows potent cytotoxic activity.

Specifications

CAS 87-44-5
Density 0.902 g/mL at 20°C
Boiling Point 262°C to 264°C
Molecular Formula C15H24
Refractive Index n20/D 1.499
Linear Formula C15H24
MDL Number MFCD00075925
Quantity 25 mL
Synonym beta-Caryophyllene; trans-(1R,9S)-8-Methylene-4,11,11-trimethylbicyclo[7.2.0]undec-4-ene
Molecular Weight (g/mol) 204.35 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.