Learn More
Medchemexpress LLC Trans-ccc_R08 | 2413192-49-9 | 99.7% | 414.84 | 25 MG
SDP
Catalog No. 5000363550
Encompass_Preferred
Trans-ccc_R08 (Compound 1-B) is a cccDNA inhibitor with anti-HBV activity. It has an IC50 of 0.14 μM for HBeAg and an IC50 of 0.08 μM for HBsAg in in vitro assays. This compound inhibits covalently closed circular DNA (cccDNA) and is applicable to research related to hepatitis B virus infection.
  • Inhibits HBeAg production in HepDES19 cells with an IC50 of 0.14 μM.
  • Inhibits HBsAg production in cryopreserved primary hepatocytes with an IC50 of 0.08 μM.
Catalog No. 50-003-63550 Supplier Medchemexpress LLC Supplier No. 25MGHY148560A
Trans-ccc_R08 (Compound 1-B) is a cccDNA inhibitor with anti-HBV activity. It has an IC50 of 0.14 μM for HBeAg and an IC50 of 0.08 μM for HBsAg in in vitro assays. This compound inhibits covalently closed circular DNA (cccDNA) and is applicable to research related to hepatitis B virus infection.
  • Inhibits HBeAg production in HepDES19 cells with an IC50 of 0.14 μM.
  • Inhibits HBsAg production in cryopreserved primary hepatocytes with an IC50 of 0.08 μM.