Learn More
Medchemexpress LLC Trans-ccc R08 | 2413192-49-9 | 99.7% | 414.84 | 5 MG
SDP
Catalog No. 5000361103
Encompass_Preferred
This cccDNA inhibitor (Compound 1-B) exhibits anti-HBV activity. It has an IC50 of 0.14 μM for HBeAg and an IC50 of 0.08 μM for HBsAg in in vitro assays. It inhibits covalently closed circular DNA (cccDNA) and is applicable to research related to hepatitis B virus infection.
  • CccDNA inhibitor
  • Anti-HBV activity
  • Inhibits HBeAg production in HepDES19 cells with an IC50 of 0.14 μM
  • Inhibits HBsAg production in cryopreserved primary hepatocytes, with an IC50 of 0.08 μM
  • Applicable to research related to hepatitis B virus infection
Catalog No. 50-003-61103 Supplier Medchemexpress LLC Supplier No. HY148560A5MG
This cccDNA inhibitor (Compound 1-B) exhibits anti-HBV activity. It has an IC50 of 0.14 μM for HBeAg and an IC50 of 0.08 μM for HBsAg in in vitro assays. It inhibits covalently closed circular DNA (cccDNA) and is applicable to research related to hepatitis B virus infection.
  • CccDNA inhibitor
  • Anti-HBV activity
  • Inhibits HBeAg production in HepDES19 cells with an IC50 of 0.14 μM
  • Inhibits HBsAg production in cryopreserved primary hepatocytes, with an IC50 of 0.08 μM
  • Applicable to research related to hepatitis B virus infection