Learn More
Apexbio Technology LLC trans-Chalcone 614-47-1 5g
SDP
Catalog No. 5000093575
Encompass_Preferred
trans-Chalcone (CAS 614-47-1) is a small-molecule inhibitor targeting fatty acid synthase (FAS) and -amylase It is designed to inhibit enzymatic activity thereby regulating lipid metabolism and carbohydrate processing trans-Chalcone exerts its biological activity primarily through inhibition of FAS and -amylase In cell-based studies trans-Chalcone demonstrates induction of cell cycle arrest and apoptosis in MCF-7 breast cancer cells Based on these pharmacological properties trans-Chalcone holds research potential in oncology enzymatic inhibition studies and cell-cycle regulation analysis
Catalog No. 50-000-93575 Supplier Apexbio Technology LLC Supplier No. A47735000
trans-Chalcone (CAS 614-47-1) is a small-molecule inhibitor targeting fatty acid synthase (FAS) and -amylase It is designed to inhibit enzymatic activity thereby regulating lipid metabolism and carbohydrate processing trans-Chalcone exerts its biological activity primarily through inhibition of FAS and -amylase In cell-based studies trans-Chalcone demonstrates induction of cell cycle arrest and apoptosis in MCF-7 breast cancer cells Based on these pharmacological properties trans-Chalcone holds research potential in oncology enzymatic inhibition studies and cell-cycle regulation analysis