Learn More
Medchemexpress LLC Trans-chalcone | 614-47-1 | MFCD00003082 | 98.8% | 208.26 g/mol | C15H12O | 5g
SDP
Catalog No. 5000268963
Encompass_Preferred
trans-Chalcone isolated from Aronia melanocarpa skin is a biphenolic core structure of flavonoids precursor trans-Chalcone is a potent fatty acid synthase (FAS) and -amylase inhibitor trans-Chalcone causes cellcycle arrest and induces apoptosis in the breastcancer cell line MCF-7 trans-Chalcone has antifungal and anticancer activity[1][2][3]
Catalog No. 50-002-68963 Supplier Medchemexpress LLC Supplier No. HYY05985G
trans-Chalcone isolated from Aronia melanocarpa skin is a biphenolic core structure of flavonoids precursor trans-Chalcone is a potent fatty acid synthase (FAS) and -amylase inhibitor trans-Chalcone causes cellcycle arrest and induces apoptosis in the breastcancer cell line MCF-7 trans-Chalcone has antifungal and anticancer activity[1][2][3]