Learn More
American Radiolabeled Chemicals Inc TRANS CINN AC 7-14C 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARC 345050UC
Encompass_Preferred
trans-Cinnamic acid,-7-14C 50uCi in ethanol, 0.1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 148.16, Exclusive
Catalog No. 50-901-263