Learn More
Apexbio Technology LLC trans-Cinnamaldehyde 14371-10-9 20mg
SDP
Catalog No. 5000090699
Encompass_Preferred
trans-Cinnamaldehyde (14371-10-9) is a small-molecule modulator applied in biomedical research to explore mechanisms involved in inflammatory responses oxidative stress modulation microbial growth inhibition and transcriptional regulation trans-Cinnamaldehyde exerts its biological activity primarily through modulation of cellular signaling pathways attributable to its -unsaturated aldehydic structure with a conjugated double bond system Based on these pharmacological properties trans-Cinnamaldehyde holds research potential in the fields of inflammation oxidative stress antimicrobial coating development and cellular signaling studies
Catalog No. 50-000-90699 Supplier Apexbio Technology LLC Supplier No. N148220
trans-Cinnamaldehyde (14371-10-9) is a small-molecule modulator applied in biomedical research to explore mechanisms involved in inflammatory responses oxidative stress modulation microbial growth inhibition and transcriptional regulation trans-Cinnamaldehyde exerts its biological activity primarily through modulation of cellular signaling pathways attributable to its -unsaturated aldehydic structure with a conjugated double bond system Based on these pharmacological properties trans-Cinnamaldehyde holds research potential in the fields of inflammation oxidative stress antimicrobial coating development and cellular signaling studies