Learn More
Medchemexpress LLC Trans-cinnamaldehyde | 14371-10-9 | MFCD00007000 | 98.0% | 132.16 g/mol | C9H8O | 5 G
SDP
Catalog No. 5000300285
Encompass_Preferred
trans-Cinnamaldehyde is an aromatic α,β-unsaturated aldehyde used in research and organic synthesis. It functions as a versatile building block for constructing substituted heterocycles, has documented antimicrobial properties, and is reported to modulate TRPA1 in cellular assays. Intended for laboratory use only; handle and store under nitrogen at recommended temperatures.
Catalog No. 50-003-00285 Supplier Medchemexpress LLC Supplier No. HYW0197115G
trans-Cinnamaldehyde is an aromatic α,β-unsaturated aldehyde used in research and organic synthesis. It functions as a versatile building block for constructing substituted heterocycles, has documented antimicrobial properties, and is reported to modulate TRPA1 in cellular assays. Intended for laboratory use only; handle and store under nitrogen at recommended temperatures.