trans-Cinnamaldehyde, ≥98%, Alfa Aesar™

Manufacturer: Alfa Aesar A1468936

Quantity 500g

 View more versions of this product

Catalog No. AAA1468936

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14371-10-9
    Physical Form Liquid
    Assay Percent Range ≥98%
    Molecular Formula C9H8O
    Formula Weight 132.16
    Chemical Name or Material trans-Cinnamaldehyde

    Specifications

    Formula Weight 132.16
    Quantity 500g
    MDL Number MFCD00007000
    Beilstein 1071571
    Merck Index 14,2297
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H317
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P280g-P262-P305+P351+P338
    Wear protective gloves.
    Do not get in eyes, on skin, or on clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue r
    RTECS Number GD6475000
    Physical Form Liquid
    Density 1.050
    Assay Percent Range ≥98%
    Molecular Formula C9H8O
    CAS 14371-10-9
    Storage Note 1 Air sensitive
    Chemical Name or Material trans-Cinnamaldehyde
    Assay ≥98%

    Description

    Applications
    trans-Cinnamaldehyde is used in the flavor and perfume industry. It is also used in medicine. It reacts with glutathione to get an adduct 1 ft.-(glutathion-S-yl)-dihydrocinnamaldehyde. It is used to prepare cinnamylidene-bisacetamide by reacting with acetamide. Further, it inhibits xanthine oxidase.

    Solubility
    Miscible with water.

    Notes
    Store in a cool place. Incompatible with strong oxidizing agents and strong bases. Air sensitive.

    GHS H Statement
    H315-H319-H317
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P280g-P262-P305+P351+P338
    Wear protective gloves.
    Do not get in eyes, on skin, or on clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue r

    Air sensitive

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit