Learn More
Medchemexpress LLC HY-N0610 100mg Medchemexpress, trans-Cinnamic acid CAS:140-10-3 Purity:>98%
SDP
Catalog No. 501933639
Encompass_Preferred
Medchemexpress, HY-N0610 100mg trans-Cinnamic acid CAS:140-10-3 trans-Cinnamic acid is a natural antimicrobial, with minimal inhibitory concentration (MIC) of 250 μg/mL against fish pathogen A. sobria, SY-AS1 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3639 Supplier Medchemexpress LLC Supplier No. HYN0610100MG
Medchemexpress, HY-N0610 100mg trans-Cinnamic acid CAS:140-10-3 trans-Cinnamic acid is a natural antimicrobial, with minimal inhibitory concentration (MIC) of 250 μg/mL against fish pathogen A. sobria, SY-AS1 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.