Learn More
Apexbio Technology LLC trans-Cinnamic acid 140-10-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000087720
Encompass_Preferred
trans-Cinnamic acid (140-10-3) is a small-molecule inhibitor that exerts inhibitory activity against specific enzyme targets involved in inflammation oxidative stress and microbial proliferation It modulates these biological processes thereby regulating pathways associated with inflammatory response and antimicrobial defense trans-Cinnamic acid exerts its biological activity primarily through enzyme inhibition In in vitro studies trans-Cinnamic acid demonstrates inhibitory activity with reported IC50 values typically between 50 200 M depending on the experimental conditions and cell models Based on these pharmacological properties trans-Cinnamic acid holds research potential in anti-inflammatory antimicrobial antioxidant response studies and metabolic assays in tissue culture and cell-based experimental models
Catalog No. 50-000-87720 Supplier Apexbio Technology LLC Supplier No. N18775.1
trans-Cinnamic acid (140-10-3) is a small-molecule inhibitor that exerts inhibitory activity against specific enzyme targets involved in inflammation oxidative stress and microbial proliferation It modulates these biological processes thereby regulating pathways associated with inflammatory response and antimicrobial defense trans-Cinnamic acid exerts its biological activity primarily through enzyme inhibition In in vitro studies trans-Cinnamic acid demonstrates inhibitory activity with reported IC50 values typically between 50 200 M depending on the experimental conditions and cell models Based on these pharmacological properties trans-Cinnamic acid holds research potential in anti-inflammatory antimicrobial antioxidant response studies and metabolic assays in tissue culture and cell-based experimental models