Learn More
Medchemexpress LLC Trans-cinnamic acid | 140-10-3 | 148.16 | 100 MG
SDP
Catalog No. 5000388614
Encompass_Preferred
Trans-Cinnamic acid is an analytical standard intended for research and analytical applications. It is a natural antimicrobial and is utilized in various qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS. This compound serves as a laboratory chemical and for the manufacture of substances.
  • Functions as an analytical standard
  • Suitable for research and analytical applications
  • Acts as a natural antimicrobial
  • Applicable in HPLC, GC, and MS experiments
  • Used as a laboratory chemical
  • Supports the manufacture of substances
Catalog No. 50-003-88614 Supplier Medchemexpress LLC Supplier No. HYN0610R100MG
Trans-Cinnamic acid is an analytical standard intended for research and analytical applications. It is a natural antimicrobial and is utilized in various qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS. This compound serves as a laboratory chemical and for the manufacture of substances.
  • Functions as an analytical standard
  • Suitable for research and analytical applications
  • Acts as a natural antimicrobial
  • Applicable in HPLC, GC, and MS experiments
  • Used as a laboratory chemical
  • Supports the manufacture of substances