Learn More
Medchemexpress LLC Trans-cinnamic acid | 140-10-3 | 148.16 | 5 G
SDP
Catalog No. 5000389033
Encompass_Preferred
Trans-Cinnamic acid is a natural antimicrobial, effective against various fish pathogens such as *A. sobria*, *A. sobria* SY-AS3, *S. baltica*, *F. spartansii* SY-FS1, *A. salmonicida* ATCC 33658, *Listonella anguillarum*, *V. crassostreae* SY-VC10, and *Y. ruckeri* E42. Its antimicrobial effectiveness is enhanced in non-neutralized culture media.
  • Natural antimicrobial properties.
  • Effective against a range of fish pathogens.
  • Antimicrobial activity is increased when the pH of the culture media is not neutralized.
Catalog No. 50-003-89033 Supplier Medchemexpress LLC Supplier No. HYN06105G
Trans-Cinnamic acid is a natural antimicrobial, effective against various fish pathogens such as *A. sobria*, *A. sobria* SY-AS3, *S. baltica*, *F. spartansii* SY-FS1, *A. salmonicida* ATCC 33658, *Listonella anguillarum*, *V. crassostreae* SY-VC10, and *Y. ruckeri* E42. Its antimicrobial effectiveness is enhanced in non-neutralized culture media.
  • Natural antimicrobial properties.
  • Effective against a range of fish pathogens.
  • Antimicrobial activity is increased when the pH of the culture media is not neutralized.