Learn More
Medchemexpress LLC Trans-Cinnamic acid | 140-10-3 | 148.16 | 500 MG
SDP
Catalog No. 5000388706
Encompass_Preferred
Trans-Cinnamic acid is a natural antimicrobial that demonstrates a minimal inhibitory concentration (MIC) of 250 μg/mL against the fish pathogen A. sobria, SY-AS1.
  • Natural antimicrobial agent.
  • Exhibits moderate inhibition against various fish pathogens.
  • More effective on bacteria when the pH of the culture media is not neutralized.
  • For research use only.
Catalog No. 50-003-88706 Supplier Medchemexpress LLC Supplier No. HYN0610500MG
Trans-Cinnamic acid is a natural antimicrobial that demonstrates a minimal inhibitory concentration (MIC) of 250 μg/mL against the fish pathogen A. sobria, SY-AS1.
  • Natural antimicrobial agent.
  • Exhibits moderate inhibition against various fish pathogens.
  • More effective on bacteria when the pH of the culture media is not neutralized.
  • For research use only.