Learn More
Medchemexpress LLC Trans-Cinnamic acid | 140-10-3 | MFCD00004369 | 148.16 | 100 MG
SDP
Catalog No. 5000388619
Encompass_Preferred
trans-Cinnamic acid is a natural antimicrobial, with a minimal inhibitory concentration (MIC) of 250 μg/mL against the fish pathogen A. sobria, SY-AS1. This compound is used in various biological applications.
  • Antimicrobial activity against various fish pathogens
  • Shows moderate inhibition on rainbow trout intestinal isolates and gill isolate
  • More effective on bacteria when the pH of the culture media is not neutralized
Catalog No. 50-003-88619 Supplier Medchemexpress LLC Supplier No. HYN0610S1100MG
trans-Cinnamic acid is a natural antimicrobial, with a minimal inhibitory concentration (MIC) of 250 μg/mL against the fish pathogen A. sobria, SY-AS1. This compound is used in various biological applications.
  • Antimicrobial activity against various fish pathogens
  • Shows moderate inhibition on rainbow trout intestinal isolates and gill isolate
  • More effective on bacteria when the pH of the culture media is not neutralized