Learn More
TARGETMOL CHEMICALS INC trans-Cinnamic acid 1G
SDP
Catalog No. 502878186
Encompass_Preferred
Also available in 1 mL, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. trans-Cinnamic acid (Myricitrine) exerts anti-diabetic activity by improving glucose tolerance in vivo and by stimulating insulin secretion in vitro. Purity 99.96%
Catalog No. 50-287-8186 Supplier TARGETMOL CHEMICALS INC Supplier No. T27401G
Also available in 1 mL, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. trans-Cinnamic acid (Myricitrine) exerts anti-diabetic activity by improving glucose tolerance in vivo and by stimulating insulin secretion in vitro. Purity 99.96%