Learn More
Medchemexpress LLC Trans-cinnamic acid-d7 | 343338-31-8 | 100.0% | 155.20 | C9HD7O2 | 10 MG
SDP
Catalog No. 5000297058
Encompass_Preferred
trans-Cinnamic acid-d7 is the deuterium-labeled analogue of trans-cinnamic acid, used as an internal standard and tracer in analytical chemistry and metabolic studies. It is supplied as a high-purity white to off-white solid intended for research applications.
  • Deuterium-labeled for use as an internal standard in mass spectrometry and tracer studies.
  • High reported purity (99.98%).
  • White to off-white solid, stable under standard storage conditions.
  • Suitable for analytical and metabolic research applications.
  • Available in a 10 mg analytical-scale quantity.
Catalog No. 50-002-97058 Supplier Medchemexpress LLC Supplier No. HYN0610S110MG
trans-Cinnamic acid-d7 is the deuterium-labeled analogue of trans-cinnamic acid, used as an internal standard and tracer in analytical chemistry and metabolic studies. It is supplied as a high-purity white to off-white solid intended for research applications.
  • Deuterium-labeled for use as an internal standard in mass spectrometry and tracer studies.
  • High reported purity (99.98%).
  • White to off-white solid, stable under standard storage conditions.
  • Suitable for analytical and metabolic research applications.
  • Available in a 10 mg analytical-scale quantity.