Learn More
Medchemexpress LLC trans-Cinnamic acid-d7 | 343338-31-8 | 155.20 | 50 MG
SDP
Catalog No. 5000388586
Encompass_Preferred
trans-Cinnamic acid-d7 (also known as trans-3-Phenylacrylic acid-d7) is the deuterium-labeled form of trans-Cinnamic acid, a natural antimicrobial. This compound is used in laboratory research and can serve as a tracer or internal standard for quantitative analysis using methods such as NMR, GC-MS, or LC-MS. Deuterium substitution in drug molecules is known for its potential to influence pharmacokinetic and metabolic profiles.
  • Deuterium-labeled form of trans-Cinnamic acid.
  • Acts as a natural antimicrobial.
  • Can be used as a tracer or internal standard.
  • Useful for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Deuterium substitution may impact pharmacokinetic and metabolic profiles.
Catalog No. 50-003-88586 Supplier Medchemexpress LLC Supplier No. HYN0610S150MG
trans-Cinnamic acid-d7 (also known as trans-3-Phenylacrylic acid-d7) is the deuterium-labeled form of trans-Cinnamic acid, a natural antimicrobial. This compound is used in laboratory research and can serve as a tracer or internal standard for quantitative analysis using methods such as NMR, GC-MS, or LC-MS. Deuterium substitution in drug molecules is known for its potential to influence pharmacokinetic and metabolic profiles.
  • Deuterium-labeled form of trans-Cinnamic acid.
  • Acts as a natural antimicrobial.
  • Can be used as a tracer or internal standard.
  • Useful for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Deuterium substitution may impact pharmacokinetic and metabolic profiles.