Learn More
Sigma Aldrich Fine Chemicals Biosciences trans Cinnamic acid natura1KG
Catalog No. 502903789
Encompass
trans-Cinnamic acid is an -unsaturated aromatic acid that can be used as a flavoring agent. It is mainly used to prepare ester derivatives that are used in perfume industry. trans-Cinnamic acid is the key volatile components of cinnamon essential oil.
Catalog No. 50-290-3789 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W2288261KGK
trans-Cinnamic acid is an -unsaturated aromatic acid that can be used as a flavoring agent. It is mainly used to prepare ester derivatives that are used in perfume industry. trans-Cinnamic acid is the key volatile components of cinnamon essential oil.