Learn More
Medchemexpress LLC Trans-cinnamic acid (standard) | 140-10-3 | 148.16 | 50 MG
SDP
Catalog No. 5000388625
Encompass_Preferred
trans-Cinnamic acid (Standard) serves as an analytical standard for research and analytical applications. It acts as a natural antimicrobial and is suitable for various laboratory experiments including HPLC, GC, and MS, as a reference standard assay.
  • Analytical standard for trans-Cinnamic acid.
  • Intended for research and analytical applications.
  • Natural antimicrobial with a minimal inhibitory concentration (MIC) of 250 μg/mL against the fish pathogen A. sobria, SY-AS1.
  • Used as a reference standard assay.
  • Utilized in qualitative, quantitative, and methodological research experiments.
  • Compatible with techniques such as HPLC, GC, and MS.
Catalog No. 50-003-88625 Supplier Medchemexpress LLC Supplier No. HYN0610R50MG
trans-Cinnamic acid (Standard) serves as an analytical standard for research and analytical applications. It acts as a natural antimicrobial and is suitable for various laboratory experiments including HPLC, GC, and MS, as a reference standard assay.
  • Analytical standard for trans-Cinnamic acid.
  • Intended for research and analytical applications.
  • Natural antimicrobial with a minimal inhibitory concentration (MIC) of 250 μg/mL against the fish pathogen A. sobria, SY-AS1.
  • Used as a reference standard assay.
  • Utilized in qualitative, quantitative, and methodological research experiments.
  • Compatible with techniques such as HPLC, GC, and MS.