Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC trans-Cinnamyl alcohol | 4407-36-7 | 134.18 | 1 ML
SDP

Catalog No. 5000415412
Encompass_Preferred

trans-Cinnamyl alcohol is a trans-isomer of Cinnamyl alcohol, an active component found in chestnut flower. It acts as an inhibitor of increased PPARγ expression and demonstrates anti-obesity activity. This compound is classified as a primary metabolite within the cinnamyl alcohol organic compounds group.

  • Purity: 98.0%
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Initial source: Plants (Rosaceae)
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at concentrations ≥ 100 mg/mL (745.27 mM)
  • In vivo protocols for clear solutions: 10% DMSO, 90% (20% SBE-β-CD in Saline) or 10% DMSO, 90% Corn Oil.

Catalog No. 50-004-15412 Supplier Medchemexpress LLC Supplier No. HYN186710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

trans-Cinnamyl alcohol is a trans-isomer of Cinnamyl alcohol, an active component found in chestnut flower. It acts as an inhibitor of increased PPARγ expression and demonstrates anti-obesity activity. This compound is classified as a primary metabolite within the cinnamyl alcohol organic compounds group.

  • Purity: 98.0%
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Initial source: Plants (Rosaceae)
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at concentrations ≥ 100 mg/mL (745.27 mM)
  • In vivo protocols for clear solutions: 10% DMSO, 90% (20% SBE-β-CD in Saline) or 10% DMSO, 90% Corn Oil.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.