Learn More
Medchemexpress LLC trans-Cinnamyl alcohol | 4407-36-7 | 134.18 | 1000 G
SDP
Catalog No. 5000415017
Encompass_Preferred
trans-Cinnamyl alcohol is a trans-isomer of Cinnamyl alcohol. Cinnamyl Alcohol, an active component from chestnut flower, inhibits increased PPARγ expression, exhibiting anti-obesity activity. trans-Cinnamyl alcohol belongs to the class of organic compounds known as cinnamyl alcohols and is considered a primary metabolite.
  • Trans-isomer of cinnamyl alcohol.
  • Active component from chestnut flower.
  • Inhibits increased PPARγ expression.
  • Possesses anti-obesity activity.
  • Primary metabolite.
  • For research use only.
Catalog No. 50-004-15017 Supplier Medchemexpress LLC Supplier No. HYN18671000G
trans-Cinnamyl alcohol is a trans-isomer of Cinnamyl alcohol. Cinnamyl Alcohol, an active component from chestnut flower, inhibits increased PPARγ expression, exhibiting anti-obesity activity. trans-Cinnamyl alcohol belongs to the class of organic compounds known as cinnamyl alcohols and is considered a primary metabolite.
  • Trans-isomer of cinnamyl alcohol.
  • Active component from chestnut flower.
  • Inhibits increased PPARγ expression.
  • Possesses anti-obesity activity.
  • Primary metabolite.
  • For research use only.