Learn More
Medchemexpress LLC trans-Cinnamyl alcohol | 4407-36-7 | 134.18 | 50 G
SDP
Catalog No. 5000415913
Encompass_Preferred
trans-Cinnamyl alcohol is a trans-isomer of Cinnamyl alcohol, an active component found in chestnut flower. It acts as an inhibitor of increased PPARγ expression and exhibits anti-obesity activity. This compound belongs to the class of organic compounds known as cinnamyl alcohols and is considered a primary metabolite.
  • For research use only.
  • It is an active component from chestnut flower.
  • It inhibits increased PPARγ expression.
  • It possesses anti-obesity activity.
  • It is a primary metabolite.
Catalog No. 50-004-15913 Supplier Medchemexpress LLC Supplier No. HYN186750G
trans-Cinnamyl alcohol is a trans-isomer of Cinnamyl alcohol, an active component found in chestnut flower. It acts as an inhibitor of increased PPARγ expression and exhibits anti-obesity activity. This compound belongs to the class of organic compounds known as cinnamyl alcohols and is considered a primary metabolite.
  • For research use only.
  • It is an active component from chestnut flower.
  • It inhibits increased PPARγ expression.
  • It possesses anti-obesity activity.
  • It is a primary metabolite.