Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,cis-2,6-Nonadien-1-ol 97.0+%, TCI America™
SDP

Catalog No. N05935ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05935ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N05935ML Supplier TCI America Supplier No. N05935ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28069-72-9
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.226
MDL Number MFCD00014055
InChI Key AMXYRHBJZOVHOL-ODYTWBPASA-N
PubChem CID 5362833
IUPAC Name (2E,6Z)-nona-2,6-dien-1-ol
SMILES CCC=CCCC=CCO

Specifications

Color Yellow
Boiling Point 100°C
Quantity 5 mL
Formula Weight 140.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material trans,cis-2,6-Nonadien-1-ol
RTECSNumber RA5394000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.