Learn More
Medchemexpress LLC Trans-Clopenthixol | 53772-84-2 | 99.4% | C22H25ClN2OS | 5 MG
SDP
Catalog No. 5000433403
Encompass_Preferred
Trans-Clopenthixol is an antibiotic (E)-isomer that does not exhibit a neuroleptic effect. It has been shown to inhibit the growth of *Pseudomonas aeruginosa* and *Plasmodium falciparum* in vitro.
  • Antibiotic without neuroleptic effect
  • Inhibits *Pseudomonas aeruginosa* in vitro
  • Inhibits *Plasmodium falciparum* in vitro
Catalog No. 50-004-33403 Supplier Medchemexpress LLC Supplier No. HYA0163D5MG
Trans-Clopenthixol is an antibiotic (E)-isomer that does not exhibit a neuroleptic effect. It has been shown to inhibit the growth of *Pseudomonas aeruginosa* and *Plasmodium falciparum* in vitro.
  • Antibiotic without neuroleptic effect
  • Inhibits *Pseudomonas aeruginosa* in vitro
  • Inhibits *Plasmodium falciparum* in vitro