Learn More
Medchemexpress LLC trans-Cyclohexane-1,2-diol | 1460-57-7 | MFCD00066430 | 99.9% | 116.16 | 10 G
SDP
Catalog No. 5000452303
Encompass_Preferred
trans-Cyclohexane-1,2-diol (TCHD) acts as a transient dilator of the nuclear pore complex (NPC). It interacts with the hydrophobic core (FG nucleoporin) of the NPC, which disrupts the NPC structure and reversibly increases the nuclear pore's permeability. This allows macromolecules larger than 40 kDa (such as plasmid DNA) to enter the cell nucleus via passive diffusion, thereby enhancing the nuclear import efficiency of non-viral vectors.
  • Can improve the efficiency of in vitro electrotransfection or lipid-mediated gene transfection.
  • Significantly increases gene expression in differentiated airway epithelial cells.
  • Allows macromolecules larger than 40 kDa to enter the cell nucleus.
  • Disrupts NPC structure and reversibly increases nuclear pore's permeability.
Catalog No. 50-004-52303 Supplier Medchemexpress LLC Supplier No. HYW01051410G
trans-Cyclohexane-1,2-diol (TCHD) acts as a transient dilator of the nuclear pore complex (NPC). It interacts with the hydrophobic core (FG nucleoporin) of the NPC, which disrupts the NPC structure and reversibly increases the nuclear pore's permeability. This allows macromolecules larger than 40 kDa (such as plasmid DNA) to enter the cell nucleus via passive diffusion, thereby enhancing the nuclear import efficiency of non-viral vectors.
  • Can improve the efficiency of in vitro electrotransfection or lipid-mediated gene transfection.
  • Significantly increases gene expression in differentiated airway epithelial cells.
  • Allows macromolecules larger than 40 kDa to enter the cell nucleus.
  • Disrupts NPC structure and reversibly increases nuclear pore's permeability.