Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trans-Dibromo[Bis(Tri-O-Tolylphosphine)]Palladium(II)

Catalog No. 7502395G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-023-95G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 75-023-95G Supplier Sigma Aldrich Supplier No. 7502395G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trans-Dibromo[Bis(Tri-O-Tolylphosphine)]Palladium(II)

Specifications

CAS 137076-54-1
Molecular Formula C28H52N4O8
Linear Formula C28H52N4O8
MDL Number MFCD02259697
Quantity 100 mg
Synonym DOTA-tri-t-Bu-ester
Molecular Weight (g/mol) 572.73 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.