Learn More
Strem, An Ascensus Company CAS# 29934-17-6. 1g. trans-Dichlorobis(tricyclohexylphosphine)palladium(II), 99%. MFCD00191830
SDP
Catalog No. 5090112346
Encompass
CAS# 29934-17-6. 1g. trans-Dichlorobis(tricyclohexylphosphine)palladium(II), 99%. MFCD00191830. Molecular Weight: 738.19. Molecular Formula: PdCl2[(C6H11)3P]2. Color/form: light yellow pwdr. Strem# 46-0520. http://www.strem.com/catalog/v/46-0520/
Catalog No. 50-901-12346 Supplier Strem, An Ascensus Company Supplier No. 4605201G
CAS# 29934-17-6. 1g. trans-Dichlorobis(tricyclohexylphosphine)palladium(II), 99%. MFCD00191830. Molecular Weight: 738.19. Molecular Formula: PdCl2[(C6H11)3P]2. Color/form: light yellow pwdr. Strem# 46-0520. http://www.strem.com/catalog/v/46-0520/