Learn More
Medchemexpress LLC Trans-dichlorobis(tricyclohexylphosphine)palladium(II) | 29934-17-6 | MFCD00191830 | 99.0% | 738.19 g·mol⁻¹ | C36H66Cl2P2Pd | 1 G
SDP
Catalog No. 5000300086
Encompass_Preferred
Dichlorobis(tricyclohexylphosphine)palladium(II) is a palladium(II) complex supplied as a light-yellow powder. It is used as a homogeneous catalyst in synthetic organic chemistry for cross-coupling and carbonylation reactions, providing reliable activity and compatibility with diverse substrates.
  • High purity (99%) for reproducible catalytic performance.
  • Light-yellow powder suitable for storage and handling.
  • Effective catalyst for C-C and C-N cross-coupling reactions.
  • Soluble in common organic solvents for homogeneous reactions.
  • Stable under typical reaction conditions used in synthesis.
Catalog No. 50-003-00086 Supplier Medchemexpress LLC Supplier No. HYW0333121G
Dichlorobis(tricyclohexylphosphine)palladium(II) is a palladium(II) complex supplied as a light-yellow powder. It is used as a homogeneous catalyst in synthetic organic chemistry for cross-coupling and carbonylation reactions, providing reliable activity and compatibility with diverse substrates.
  • High purity (99%) for reproducible catalytic performance.
  • Light-yellow powder suitable for storage and handling.
  • Effective catalyst for C-C and C-N cross-coupling reactions.
  • Soluble in common organic solvents for homogeneous reactions.
  • Stable under typical reaction conditions used in synthesis.