Learn More
Strem, An Ascensus Company CAS# 13965-03-2. 5g. trans-Dichlorobis(triphenylphosphine)palladium(II), 99% (99.9+%-Pd). MFCD00009593
SDP
Catalog No. 5090112351
Encompass
CAS# 13965-03-2. 5g. trans-Dichlorobis(triphenylphosphine)palladium(II), 99% (99.9+%-Pd). MFCD00009593. Molecular Weight: 701.89. Molecular Formula: PdCl2(P(C6H5)3)2. Color/form: yellow pwdr. Strem# 46-0530. http://www.strem.com/catalog/v/46-0530/
Catalog No. 50-901-12351 Supplier Strem, An Ascensus Company Supplier No. 4605305G
CAS# 13965-03-2. 5g. trans-Dichlorobis(triphenylphosphine)palladium(II), 99% (99.9+%-Pd). MFCD00009593. Molecular Weight: 701.89. Molecular Formula: PdCl2(P(C6H5)3)2. Color/form: yellow pwdr. Strem# 46-0530. http://www.strem.com/catalog/v/46-0530/