Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trans-Dichlorobis(Triphenylphosphine)Platinumf(II)

Catalog No. 2884031G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-840-31G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 28-840-31G Supplier Sigma Aldrich Supplier No. 2884031G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Catalyst for:Preparation of soluble polymers of monosubstituted acetylenes with quaternary ammonium pendant groupsAlkynylation and hydrogenation reactionsHetero-Diels-Alder/ring-opening reactionsInsertion of carbenes into O-H bonds of alcoholsCyclopropanation of olefinsHydroformylation reactions

Specifications

CAS 18643-08-8
Melting Point 28°C to 30°C
Boiling Point 145°C to 155°C (0.005 mmHg, lit.)
Molecular Formula C20H43ClSi
Linear Formula CH3(CH2)17Si(CH3)2Cl
MDL Number MFCD00000504
Quantity 25 g
Synonym trans-Bis(triphenylphosphine)platinum dichloride
Molecular Weight (g/mol) 347.09 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.