Learn More
Medchemexpress LLC HY-75992 1mg Medchemexpress, trans-Doxercalciferol CAS:74007-20-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY759921MG
Encompass_Preferred
Medchemexpress, HY-75992 1mg trans-Doxercalciferol CAS:74007-20-8 trans-Doxercalciferol is an isomer of Doxercalciferol. Doxercalciferol is a Vitamin D2 analog, acts as an activator of Vitamin D receptor, and prevent renal disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0079