Learn More
Medchemexpress LLC Trans-doxercalciferol | 74007-20-8 | 98.2% | C28H44O2 | 1 ML
SDP
Catalog No. 5000434672
Encompass_Preferred
trans-Doxercalciferol is an isomer of Doxercalciferol, a synthetic vitamin D2 analog. It acts as an activator of Vitamin D receptors and is used in various research applications. Store the product tightly sealed in a cool, well-ventilated area, away from direct sunlight and ignition sources. Recommended storage is -20°C for powder and -80°C for solutions in solvent.
  • An isomer of Doxercalciferol
  • Activator of Vitamin D receptors
  • Suitable for research purposes
Catalog No. 50-004-34672 Supplier Medchemexpress LLC Supplier No. HY7599210MM/1ML
trans-Doxercalciferol is an isomer of Doxercalciferol, a synthetic vitamin D2 analog. It acts as an activator of Vitamin D receptors and is used in various research applications. Store the product tightly sealed in a cool, well-ventilated area, away from direct sunlight and ignition sources. Recommended storage is -20°C for powder and -80°C for solutions in solvent.
  • An isomer of Doxercalciferol
  • Activator of Vitamin D receptors
  • Suitable for research purposes