Learn More
Medchemexpress LLC Trans-doxercalciferol | 74007-20-8 | MFCD01076571 | 98.2% | 412.65 | C28H44O2 | 10 MG
SDP
Catalog No. 5000298464
Encompass_Preferred
trans-Doxercalciferol is an isomer of doxercalciferol (a vitamin D2 analog) that activates the vitamin D receptor (VDR). Supplied as a research-grade powder for in vitro and analytical use, it has a molecular weight of 412.65 g/mol and a reported purity of 98.2%.
  • Is an isomer of doxercalciferol and a vitamin D2 analog.
  • Activates the vitamin D receptor (VDR) and is used in vitamin D signaling research.
  • High reported purity (98.2%).
  • Available in multiple pack sizes, including 10 mg.
  • Powder storage: -20°C for long-term, 4°C for short-term stability.
Catalog No. 50-002-98464 Supplier Medchemexpress LLC Supplier No. HY7599210MG
trans-Doxercalciferol is an isomer of doxercalciferol (a vitamin D2 analog) that activates the vitamin D receptor (VDR). Supplied as a research-grade powder for in vitro and analytical use, it has a molecular weight of 412.65 g/mol and a reported purity of 98.2%.
  • Is an isomer of doxercalciferol and a vitamin D2 analog.
  • Activates the vitamin D receptor (VDR) and is used in vitamin D signaling research.
  • High reported purity (98.2%).
  • Available in multiple pack sizes, including 10 mg.
  • Powder storage: -20°C for long-term, 4°C for short-term stability.