Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-Ferulic acid Matrix Substance for MALDI-MS (HPLC), ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012876 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2876 1 g
11-101-2877 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2876 Supplier MilliporeSigma Supelco Supplier No. 462781GF
Add to Cart
Add to Cart

Specifications

CAS 126833-17-8
Physical Form Powder
Linear Formula HOC6H3(OCH3)CH=CHCO2 H
MDL Number MFCD00004400
Melting Point 168°C to 172°C; 171°C to 175°C
Synonym trans-4-Hydroxy-3-methoxycinnamic acid; Ferulic acid
Type Matrix Substance for MALDI-MS
Quantity 1 g
Formula Weight 194.18
Percent Purity ≥99.0% (HPLC)
Grade Matrix Substance for MALDI-MS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.