Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC trans-Isoferulic acid | 25522-33-2 | 194.19 | 250 MG
SDP

Catalog No. 5000389578
Encompass_Preferred

trans-Isoferulic acid (trans-3-Hydroxy-4-methoxycinnamic acid) is an aromatic acid isolated from the roots of *Clematis florida* var. *plena*. It exhibits anti-inflammatory activity and suppresses NO and PGE2 production through the induction of Nrf2-dependent heme oxygenase-1 (HO-1).

  • Purity: 99.93%
  • Appearance: Solid
  • Color: White to light brown
  • Initial source: Plants Ranunculaceae
  • Biological activity: Suppresses NO and PGE2 production by inducing Nrf2-dependent HO-1.
  • Target: EP, Human endogenous metabolite
  • In vitro solubility: DMSO: 100 mg/mL (514.96 mM)

Catalog No. 50-003-89578 Supplier Medchemexpress LLC Supplier No. 250MGHYN0761A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

trans-Isoferulic acid (trans-3-Hydroxy-4-methoxycinnamic acid) is an aromatic acid isolated from the roots of *Clematis florida* var. *plena*. It exhibits anti-inflammatory activity and suppresses NO and PGE2 production through the induction of Nrf2-dependent heme oxygenase-1 (HO-1).

  • Purity: 99.93%
  • Appearance: Solid
  • Color: White to light brown
  • Initial source: Plants Ranunculaceae
  • Biological activity: Suppresses NO and PGE2 production by inducing Nrf2-dependent HO-1.
  • Target: EP, Human endogenous metabolite
  • In vitro solubility: DMSO: 100 mg/mL (514.96 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.