Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC trans-Melilotoside | 618-67-7 | 326.30 | 1 MG
SDP

Catalog No. 5000414785
Encompass_Preferred

trans-Melilotoside is a natural product isolated from *Mikania laevigata*, intended exclusively for research use. It exhibits various cellular effects, including cytotoxicity against multiple cancer cell lines, making it a valuable compound for scientific investigation.

  • High purity (99.79%) for reliable experimental results
  • Active against A549, B16, BGC-823, Bel-7402, DU-145, HT-29, MCF7, MDA-MB-231, and SGC-7901 cell lines
  • Isolated from a natural plant source

Catalog No. 50-004-14785 Supplier Medchemexpress LLC Supplier No. HYN11173A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

trans-Melilotoside is a natural product isolated from *Mikania laevigata*, intended exclusively for research use. It exhibits various cellular effects, including cytotoxicity against multiple cancer cell lines, making it a valuable compound for scientific investigation.

  • High purity (99.79%) for reliable experimental results
  • Active against A549, B16, BGC-823, Bel-7402, DU-145, HT-29, MCF7, MDA-MB-231, and SGC-7901 cell lines
  • Isolated from a natural plant source

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.