Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC trans-Melilotoside | 618-67-7 | 326.30 g/mol | 5 MG
SDP

Catalog No. 5000414824
Encompass_Preferred

trans-Melilotoside (compound 3) is a natural product that can be isolated from Mikania laevigata. It is intended for research use only and is not sold to patients.

  • Exhibits cytotoxicity against various cell lines, including A549, B16, BGC-823, Bel-7402, DU-145, HT-29, MCF7, MDA-MB-231, and SGC-7901.
  • Has an IC50 of 1.6 μg/mL for compound 10 in cytotoxic studies.

Catalog No. 50-004-14824 Supplier Medchemexpress LLC Supplier No. HYN11173A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

trans-Melilotoside (compound 3) is a natural product that can be isolated from Mikania laevigata. It is intended for research use only and is not sold to patients.

  • Exhibits cytotoxicity against various cell lines, including A549, B16, BGC-823, Bel-7402, DU-145, HT-29, MCF7, MDA-MB-231, and SGC-7901.
  • Has an IC50 of 1.6 μg/mL for compound 10 in cytotoxic studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.