Learn More
AdipoGen trans-Methyl-2-dodecenoic acid
SDP
Catalog No. 502561042
Encompass_Preferred
Chemical. CAS 677354-24-4. Formula C13H24O2. MW 212.3. Synthetic. trans-Delta2-11-methyl-Dodecenoic acid is the conformational isomer of cis-Delta2-11-methyl-dodecenoic acid, a diffusible signal factor DSF in extracellular microbial and fungal communication systems. Both prokaryotes and eukaryotes depend on small signalling molecules for cell-cell communication. In a DSF bioassay, the minimum concentration of cis-Delta2-11-methyl-dodecenoic acid required for induction of a DSF biosensor was about 0.5 µM, which is 200-fold lower than that of trans-Delta2-11-methyl-dodecenoic acid and 20,000-fold lower than that of the corresponding saturated fatty acid 11-methyl-dodecanoic acid.
Catalog No. 50-256-1042 Supplier AdipoGen Supplier No. CDXD0791M025
Add to Cart
Add to Cart
Chemical. CAS 677354-24-4. Formula C13H24O2. MW 212.3. Synthetic. trans-Delta2-11-methyl-Dodecenoic acid is the conformational isomer of cis-Delta2-11-methyl-dodecenoic acid, a diffusible signal factor DSF in extracellular microbial and fungal communication systems. Both prokaryotes and eukaryotes depend on small signalling molecules for cell-cell communication. In a DSF bioassay, the minimum concentration of cis-Delta2-11-methyl-dodecenoic acid required for induction of a DSF biosensor was about 0.5 µM, which is 200-fold lower than that of trans-Delta2-11-methyl-dodecenoic acid and 20,000-fold lower than that of the corresponding saturated fatty acid 11-methyl-dodecanoic acid.