Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-β-Methylstyrene, Stabilized 99%, Thermo Scientific™

Catalog No. AC127720250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC127720250 25 g
AC127720050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC127720250 Supplier Thermo Scientific Chemicals Supplier No. 127720250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-50-3
Molecular Formula C9H10
Molecular Weight (g/mol) 118.18
MDL Number MFCD00009280
InChI Key QROGIFZRVHSFLM-QHHAFSJGSA-N
Synonym beta-methylstyrene, trans-beta-methylstyrene, 1-phenylpropene, isoallylbenzene, propenylbenzene, trans-1-phenyl-1-propene, 1-propenylbenzene, trans-propenylbenzene, 1-phenyl-1-propene, benzene, 1-propenyl
PubChem CID 252325
IUPAC Name [(1E)-prop-1-en-1-yl]benzene
SMILES C\C=C\C1=CC=CC=C1

Specifications

Color Colorless to Yellow
Density 0.91
Infrared Spectrum Authentic
Linear Formula C6H5CH=CHCH3
Refractive Index 1.5480 to 1.5500 (20°C, 589nm)
Quantity 25 g
Beilstein 05, 481
Stabilizer 0.01% BHT
Formula Weight 118.18
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material trans-β-Methylstyrene, 99%
Show More Show Less
EINECSNumber 211-287-9
RTECSNumber DA8400000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.