Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-N-Carbobenzoxy-4-hydroxy-L-proline 98.0+%, TCI America™
SDP

Catalog No. C25065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25065G 5 g
C250625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25065G Supplier TCI America Supplier No. C25065G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13504-85-3
Molecular Formula C13H15NO5
Molecular Weight (g/mol) 265.27
MDL Number MFCD00037329,MFCD00037329,MFCD00238423,MFCD00037329,MFCD00238423,MFCD03428474
InChI Key WWVCWLBEARZMAH-UHFFFAOYNA-N
Synonym 2s-1-benzyloxy carbonyl-4-hydroxypyrrolidine-2-carboxylic acid, z-l-4-hydroxyproline, n-cbz-4-hydroxyproline, n-cbz-4-hydroxy-l-proline, n-benzyloxycarbonyl-4-hydroxyproline, n-carbobenzyloxy-4-hydroxy-l-proline, 4-hydroxy-n-benzyloxycarbonyl-l-proline, 1-carbobenzyloxy-4-hydroxy-l-proline, s-1-cbz-4-hydroxy-2-pyrrolidinecarboxylic acid
PubChem CID 12731799
IUPAC Name 1-[(benzyloxy)carbonyl]-4-hydroxypyrrolidine-2-carboxylic acid
SMILES OC1CC(N(C1)C(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 107°C
Color White
Quantity 5 g
Formula Weight 265.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Lumps
Chemical Name or Material trans-N-Carbobenzoxy-4-hydroxy-L-proline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.