Learn More
Medchemexpress LLC Trans-N,N-bis[2-chlorophenylmethyl]-1,4-cyclohexanedimethanamine dihydrochloride | 366-93-8 | MFCD01734586 | 99.7% | 464.30 g/mol | C22H30Cl4N2 | 10 MG
SDP
Catalog No. 5000239898
Encompass_Preferred
AY 9944 dihydrochloride is a research-grade small molecule that selectively inhibits 7-dehydrocholesterol Δ7-reductase (DHCR7), disrupting cholesterol biosynthesis for biochemical and cellular studies. Supplied as a solid dihydrochloride salt with high purity, it is intended for laboratory research use where potent, reproducible DHCR7 inhibition is required.
  • Inhibits DHCR7 with an IC50 of 13 nM.
  • High purity (~99.7%) suitable for analytical and biological assays.
  • Solid dihydrochloride form for convenient handling and weighing.
  • Soluble in DMSO and water; sonication may be required.
  • Stable when stored sealed at 4°C; stock solutions stable at -20°C to -80°C.
  • Commonly used to model cholesterol biosynthesis inhibition in vitro and in vivo.
Catalog No. 50-002-39898 Supplier Medchemexpress LLC Supplier No. HY10742010MG
AY 9944 dihydrochloride is a research-grade small molecule that selectively inhibits 7-dehydrocholesterol Δ7-reductase (DHCR7), disrupting cholesterol biosynthesis for biochemical and cellular studies. Supplied as a solid dihydrochloride salt with high purity, it is intended for laboratory research use where potent, reproducible DHCR7 inhibition is required.
  • Inhibits DHCR7 with an IC50 of 13 nM.
  • High purity (~99.7%) suitable for analytical and biological assays.
  • Solid dihydrochloride form for convenient handling and weighing.
  • Soluble in DMSO and water; sonication may be required.
  • Stable when stored sealed at 4°C; stock solutions stable at -20°C to -80°C.
  • Commonly used to model cholesterol biosynthesis inhibition in vitro and in vivo.